siRNA Details
| Gene Symbol: Casq2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017131.2 | NP_058827.3 | 11 | 1339 | ||
| GenBank mRNA | BC072547.1 | - | 11 | 1326 | 7105911 | |
| U33287.1 | - | 11 | 1320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Casq2| Gene Symbol: Casq2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017131.2 | NP_058827.3 | 11 | 1339 | ||
| GenBank mRNA | BC072547.1 | - | 11 | 1326 | 7105911 | |
| U33287.1 | - | 11 | 1320 | |||