siRNA Details
| Gene Symbol: Sqle | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017136.2 | NP_058832.1 | 2 | 410 | ||
| GenBank mRNA | BC097330.1 | - | 2 | 410 | 7460860 | |
| D37920.1 | - | 2 | 541 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sqle| Gene Symbol: Sqle | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017136.2 | NP_058832.1 | 2 | 410 | ||
| GenBank mRNA | BC097330.1 | - | 2 | 410 | 7460860 | |
| D37920.1 | - | 2 | 541 | |||