siRNA Details
| Gene Symbol: GCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012198.3 | NP_036330.1 | 5 | 559 | ||
| XM_005246446.2 | XP_005246503.1 | 5 | 500 | |||
| XM_006712398.3 | XP_006712461.1 | 5 | 732 | |||
| XM_006712400.3 | XP_006712463.1 | 6 | 810 | |||
| XM_006712401.3 | XP_006712464.1 | 5 | 811 | |||
| XM_011510926.2 | XP_011509228.1 | 5 | 511 | |||
| XM_011510927.2 | XP_011509229.1 | 5 | 506 | |||
| XM_011510928.2 | XP_011509230.1 | 3 | 444 | |||
| XM_017003764.1 | XP_016859253.1 | 6 | 823 | |||
| XM_017003765.1 | XP_016859254.1 | 6 | 909 | |||
| XM_017003766.1 | XP_016859255.1 | 6 | 706 | |||
| XM_017003767.1 | XP_016859256.1 | 5 | 582 | |||
| XM_017003768.1 | XP_016859257.1 | 5 | 577 | |||
| GenBank mRNA | AB209768.1 | - | 5 | 448 | ||
| AK295413.1 | - | 5 | 492 | |||
| AK312349.1 | - | 5 | 508 | |||
| BC005214.1 | - | 5 | 482 | 3996889 | ||
| M81637.1 | - | 5 | 502 | |||

GCA
Genomic Map