siRNA Details
| Gene Symbol: Gif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017162.3 | NP_058858.1 | 2 | 250 | ||
| GenBank mRNA | J03577.1 | - | 2 | 251 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gif| Gene Symbol: Gif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017162.3 | NP_058858.1 | 2 | 250 | ||
| GenBank mRNA | J03577.1 | - | 2 | 251 | ||