siRNA Details
| Gene Symbol: Fah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017181.2 | NP_058877.1 | 13 | 1212 | ||
| GenBank mRNA | BC076381.1 | - | 13 | 1212 | 7108528 | |
| FQ219602.1 | - | 13 | 1171 | |||
| M77694.1 | - | 13 | 1134 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fah| Gene Symbol: Fah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017181.2 | NP_058877.1 | 13 | 1212 | ||
| GenBank mRNA | BC076381.1 | - | 13 | 1212 | 7108528 | |
| FQ219602.1 | - | 13 | 1171 | |||
| M77694.1 | - | 13 | 1134 | |||