siRNA Details
| Gene Symbol: Gcm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017186.2 | NP_058882.1 | 5, 6 | 736 | ||
| GenBank mRNA | AF081557.1 | - | 5, 6 | 737 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gcm1| Gene Symbol: Gcm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017186.2 | NP_058882.1 | 5, 6 | 736 | ||
| GenBank mRNA | AF081557.1 | - | 5, 6 | 737 | ||