siRNA Details
| Gene Symbol: Mag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017190.4 | NP_058886.1 | 11 | 1869 | ||
| XM_006228824.2 | XP_006228886.1 | 10 | 1899 | |||
| XM_006228825.2 | XP_006228887.1 | 10 | 2416 | |||
| XM_008759137.1 | XP_008757359.1 | 10 | 2423 | |||
| XM_008759138.1 | XP_008757360.1 | 10 | 1892 | |||
| GenBank mRNA | M11721.1 | - | 5 | 785 | ||
| M14871.1 | - | 10 | 1839 | |||
| M16800.1 | - | 11 | 1869 | |||
| M36702.1 | - | 5 | 785 | |||
| X05301.1 | - | 10 | 1832 | |||

Mag
Genomic Map