siRNA Details
| Gene Symbol: Gap43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017195.3 | NP_058891.1 | 2 | 303 | ||
| GenBank mRNA | BC127502.1 | - | 2 | 151 | 8364847 | |
| FQ214107.1 | - | 2 | 138 | |||
| M16228.1 | - | 2 | 156 | |||
| M16736.1 | - | 2 | 139 | |||
| X06338.1 | - | 2 | 303 | |||

Gap43
Genomic Map