siRNA Details
| Gene Symbol: Avpr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017205.3 | NP_058901.3 | 2 | 1264 | ||
| XM_008769445.1 | XP_008767667.1 | 1 | 924 | |||
| GenBank mRNA | D45400.1 | - | 1 | 781 | ||
| U27322.1 | - | 1 | 1157 | |||
| Gene Symbol: LOC100909648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289800.1 | NP_001276729.1 | 1 | 732 | ||
| XM_008769404.1 | XP_008767626.1 | 1 | 917 | |||
More Information
Set Membership: |
|

Avpr1b
Genomic Map