siRNA Details
| Gene Symbol: Slc6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017206.1 | NP_058902.1 | 10, 11 | 1339 | ||
| XM_006236903.2 | XP_006236965.1 | 9, 10 | 1448 | |||
| XM_008763123.1 | XP_008761345.1 | 11, 12 | 1609 | |||
| GenBank mRNA | M96601.1 | - | 10, 11 | 1339 | ||

Slc6a6
Genomic Map