siRNA Details
| Gene Symbol: Lbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017208.2 | NP_058904.2 | 4 | 508 | ||
| GenBank mRNA | BC101906.1 | - | 4 | 508 | 7933709 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lbp| Gene Symbol: Lbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017208.2 | NP_058904.2 | 4 | 508 | ||
| GenBank mRNA | BC101906.1 | - | 4 | 508 | 7933709 | |