siRNA Details
| Gene Symbol: Mapt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017212.2 | NP_058908.2 | 2, 3 | 239 | ||
| XM_008768271.1 | XP_008766493.1 | 2, 3 | 278 | |||
| XM_008768281.1 | XP_008766503.1 | 2, 3 | 271 | |||
| GenBank mRNA | BC126095.1 | - | 2, 3 | 239 | 8367931 | |
| CB581821.1 | - | 2, 3 | 219 | |||
| DY311887.1 | - | 2, 3 | 239 | |||
| X79321.1 | - | 2, 3 | 326 | |||

Mapt
Genomic Map