siRNA Details
| Gene Symbol: Rgs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017214.1 | NP_058910.1 | 3, 4 | 321 | ||
| GenBank mRNA | AF117211.1 | - | 3, 4 | 321 | ||
| FQ215804.1 | - | 3, 4 | 267 | |||
| U27767.1 | - | 3, 4 | 275 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rgs4| Gene Symbol: Rgs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017214.1 | NP_058910.1 | 3, 4 | 321 | ||
| GenBank mRNA | AF117211.1 | - | 3, 4 | 321 | ||
| FQ215804.1 | - | 3, 4 | 267 | |||
| U27767.1 | - | 3, 4 | 275 | |||