siRNA Details
| Gene Symbol: Slc1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035233.1 | NP_001030310.1 | 251 | |||
| NM_017215.2 | NP_058911.2 | 251 | ||||
| GenBank mRNA | AF297648.1 | - | 163 | |||
| AF465909.1 | - | 144 | ||||
| AY044832.1 | - | 140 | ||||
| CB760876.1 | - | 252 | ||||
| JN132400.1 | - | 545 | ||||
| U15098.1 | - | 200 | ||||
| X67857.1 | - | 238 | ||||

Slc1a2
Genomic Map