siRNA Details
| Gene Symbol: Slc3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017216.1 | NP_058912.1 | 1 | 398 | ||
| GenBank mRNA | BC078852.1 | - | 1 | 393 | 7110514 | |
| M77345.1 | - | 1 | 398 | |||
| M80804.1 | - | 1 | 424 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc3a1| Gene Symbol: Slc3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017216.1 | NP_058912.1 | 1 | 398 | ||
| GenBank mRNA | BC078852.1 | - | 1 | 393 | 7110514 | |
| M77345.1 | - | 1 | 398 | |||
| M80804.1 | - | 1 | 424 | |||