siRNA Details
| Gene Symbol: Slc22a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017224.2 | NP_058920.1 | 1 | 207 | ||
| XM_006230978.2 | XP_006231040.1 | 1 | 462 | |||
| GenBank mRNA | AB004559.1 | - | 1 | 437 | ||
| AF008221.1 | - | 1 | 415 | |||
| AF110022.1 | - | 1 | 109 | |||
| BC104692.1 | - | 1 | 207 | 7109188 | ||

Slc22a6
Genomic Map