siRNA Details
| Gene Symbol: Pebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017236.1 | NP_058932.1 | 4 | 1029 | ||
| GenBank mRNA | FQ211637.1 | - | 4 | 991 | ||
| X75253.1 | - | 4 | 1029 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pebp1| Gene Symbol: Pebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017236.1 | NP_058932.1 | 4 | 1029 | ||
| GenBank mRNA | FQ211637.1 | - | 4 | 991 | ||
| X75253.1 | - | 4 | 1029 | |||