siRNA Details
| Gene Symbol: Myh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017240.2 | NP_058936.1 | 14 | 1861 | ||
| XM_006251951.2 | XP_006252013.1 | 16 | 2011 | |||
| GenBank mRNA | X15939.1 | - | 14 | 1863 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myh7| Gene Symbol: Myh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017240.2 | NP_058936.1 | 14 | 1861 | ||
| XM_006251951.2 | XP_006252013.1 | 16 | 2011 | |||
| GenBank mRNA | X15939.1 | - | 14 | 1863 | ||