siRNA Details
| Gene Symbol: Map2k5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033987.1 | NP_001029159.1 | 4 | 883 | ||
| NM_017246.2 | NP_058942.1 | 4 | 883 | |||
| XM_006243229.2 | XP_006243291.1 | 4 | 942 | |||
| XM_008766221.1 | XP_008764443.1 | 2 | 81 | |||
| GenBank mRNA | BC078860.1 | - | 4 | 883 | 7122708 | |
| U37462.1 | - | 4 | 371 | |||
| U37463.1 | - | 2 | 92 | |||
| U37464.1 | - | 4 | 371 | |||

Map2k5
Genomic Map