siRNA Details
| Gene Symbol: Htr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017254.1 | NP_058950.1 | 1 | 330 | ||
| GenBank mRNA | M30705.1 | - | 1 | 330 | ||
| M64867.1 | - | 736 | ||||
| X13971.1 | - | 873 | ||||
More Information
Set Membership: |
|

Htr2a
Genomic Map