siRNA Details
| Gene Symbol: Psme2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017257.2 | NP_058953.1 | 8, 9 | 494 | ||
| GenBank mRNA | BC058486.1 | - | 8, 9 | 532 | 6918985 | |
| BC097331.1 | - | 8, 9 | 494 | 7389180 | ||
| D45250.1 | - | 8, 9 | 544 | |||
| FQ226805.1 | - | 8, 9 | 525 | |||
| FQ227049.1 | - | 8, 9 | 538 | |||
| FQ232840.1 | - | 8, 9 | 524 | |||
| FQ233763.1 | - | 8, 9 | 520 | |||

Psme2
Genomic Map