siRNA Details
| Gene Symbol: Ptprj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017269.2 | NP_058965.2 | 3 | 417 | ||
| GenBank mRNA | U40790.1 | - | 3 | 347 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprj| Gene Symbol: Ptprj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017269.2 | NP_058965.2 | 3 | 417 | ||
| GenBank mRNA | U40790.1 | - | 3 | 347 | ||