siRNA Details
| Gene Symbol: Adh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017270.2 | NP_058966.2 | 3 | 290 | ||
| GenBank mRNA | BC127504.1 | - | 3 | 290 | 7325543 | |
| X90710.1 | - | 3 | 316 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adh4| Gene Symbol: Adh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017270.2 | NP_058966.2 | 3 | 290 | ||
| GenBank mRNA | BC127504.1 | - | 3 | 290 | 7325543 | |
| X90710.1 | - | 3 | 316 | |||