siRNA Details
| Gene Symbol: Gdi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017276.2 | NP_058972.2 | 6, 7 | 827 | ||
| GenBank mRNA | BC061767.1 | - | 6, 7 | 827 | 5621162 | |
| X74401.1 | - | 6, 7 | 757 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gdi2| Gene Symbol: Gdi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017276.2 | NP_058972.2 | 6, 7 | 827 | ||
| GenBank mRNA | BC061767.1 | - | 6, 7 | 827 | 5621162 | |
| X74401.1 | - | 6, 7 | 757 | |||