siRNA Details
| Gene Symbol: ZNF212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012256.3 | NP_036388.2 | 5 | 1852 | ||
| XM_005250055.2 | XP_005250112.2 | 5 | 1850 | |||
| XM_011516581.2 | XP_011514883.2 | 5 | 1837 | |||
| XM_011516582.1 | XP_011514884.1 | 5 | 1772 | |||
| XM_017012646.1 | XP_016868135.1 | 5 | 1850 | |||
| XM_017012647.1 | XP_016868136.1 | 6 | 1806 | |||
| XM_017012648.1 | XP_016868137.1 | 5 | 2095 | |||
| GenBank mRNA | AK074821.1 | - | 6 | 1855 | ||
| BC022785.2 | - | 5 | 1819 | 3849803 | ||
| U38864.1 | - | 3 | 1302 | |||

ZNF212
Genomic Map