siRNA Details
| Gene Symbol: Psma5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017282.1 | NP_058978.1 | 2, 3 | 188 | ||
| GenBank mRNA | BC060575.1 | - | 2, 3 | 209 | 6920479 | |
| D10756.1 | - | 2, 3 | 188 | |||
| FQ218317.1 | - | 2, 3 | 205 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psma5| Gene Symbol: Psma5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017282.1 | NP_058978.1 | 2, 3 | 188 | ||
| GenBank mRNA | BC060575.1 | - | 2, 3 | 209 | 6920479 | |
| D10756.1 | - | 2, 3 | 188 | |||
| FQ218317.1 | - | 2, 3 | 205 | |||