siRNA Details
| Gene Symbol: Psmb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017285.1 | NP_058981.1 | 4 | 432 | ||
| GenBank mRNA | BC084723.1 | - | 3 | 355 | 7319054 | |
| D21800.1 | - | 4 | 432 | |||
| FQ211229.1 | - | 4 | 389 | |||
| FQ232780.1 | - | 4 | 428 | |||

Psmb3
Genomic Map