siRNA Details
Gene Symbol: Cyp11a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017286.2 | NP_058982.1 | 3 | 566 | ||
GenBank mRNA | BC089100.1 | - | 3 | 591 | 7307104 | |
J05156.1 | - | 3 | 586 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cyp11a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017286.2 | NP_058982.1 | 3 | 566 | ||
GenBank mRNA | BC089100.1 | - | 3 | 591 | 7307104 | |
J05156.1 | - | 3 | 586 |