siRNA Details
| Gene Symbol: Gabrr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017292.2 | NP_058988.1 | 1, 2 | 284 | ||
| XM_008763610.1 | XP_008761832.1 | 3, 4 | 270 | |||
| GenBank mRNA | D38494.1 | - | 1, 2 | 284 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabrr2| Gene Symbol: Gabrr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017292.2 | NP_058988.1 | 1, 2 | 284 | ||
| XM_008763610.1 | XP_008761832.1 | 3, 4 | 270 | |||
| GenBank mRNA | D38494.1 | - | 1, 2 | 284 | ||