siRNA Details
| Gene Symbol: Pacsin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017294.1 | NP_058990.1 | 5, 6 | 750 | ||
| GenBank mRNA | AF104402.1 | - | 5, 6 | 750 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pacsin1| Gene Symbol: Pacsin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017294.1 | NP_058990.1 | 5, 6 | 750 | ||
| GenBank mRNA | AF104402.1 | - | 5, 6 | 750 | ||