siRNA Details
| Gene Symbol: Cacna1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017298.1 | NP_058994.1 | 43, 44 | 5881 | ||
| XM_006252586.2 | XP_006252648.1 | 44, 45 | 5952 | |||
| XM_006252590.2 | XP_006252652.1 | 43, 44 | 5871 | |||
| XM_006252594.2 | XP_006252656.1 | 42, 43 | 5811 | |||
| XM_006252595.2 | XP_006252657.1 | 41, 42 | 5766 | |||
| XM_008770990.1 | XP_008769212.1 | 35, 36 | 6814 | |||
| GenBank mRNA | AF370010.1 | - | 7, 8 | 1003 | ||
| D38101.1 | - | 43, 44 | 5881 | |||
| D38102.1 | - | 43, 44 | 5823 | |||

Cacna1d
Genomic Map