siRNA Details
| Gene Symbol: Eci1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017306.4 | NP_059002.2 | 4 | 396 | ||
| GenBank mRNA | BC078705.1 | - | 4 | 396 | 7129389 | |
| FQ213384.1 | - | 4 | 404 | |||
| FQ218351.1 | - | 4 | 397 | |||
| M61112.1 | - | 4 | 385 | |||
| X61184.1 | - | 4 | 407 | |||

Eci1
Genomic Map