siRNA Details
Gene Symbol: Slc25a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017307.3 | NP_059003.1 | 6 | 678 | ||
GenBank mRNA | BC100077.1 | - | 6 | 662 | 7126394 | |
FQ214871.1 | - | 3 | 938 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc25a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017307.3 | NP_059003.1 | 6 | 678 | ||
GenBank mRNA | BC100077.1 | - | 6 | 662 | 7126394 | |
FQ214871.1 | - | 3 | 938 |