siRNA Details
| Gene Symbol: Ptpro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017336.1 | NP_059032.1 | 2 | 330 | ||
| GenBank mRNA | U28938.1 | - | 2 | 330 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpro| Gene Symbol: Ptpro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017336.1 | NP_059032.1 | 2 | 330 | ||
| GenBank mRNA | U28938.1 | - | 2 | 330 | ||