siRNA Details
| Gene Symbol: Pdlim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017365.2 | NP_059061.1 | 7 | 1341 | ||
| GenBank mRNA | BC072465.1 | - | 7 | 1332 | 7100446 | |
| FQ227333.1 | - | 4 | 860 | |||
| U23769.1 | - | 7 | 1341 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdlim1| Gene Symbol: Pdlim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017365.2 | NP_059061.1 | 7 | 1341 | ||
| GenBank mRNA | BC072465.1 | - | 7 | 1332 | 7100446 | |
| FQ227333.1 | - | 4 | 860 | |||
| U23769.1 | - | 7 | 1341 | |||