siRNA Details
| Gene Symbol: Syt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019122.1 | NP_061995.1 | 5 | 1187 | ||
| XM_006228978.2 | XP_006229040.1 | 6 | 1645 | |||
| XM_006228979.2 | XP_006229041.1 | 6 | 1650 | |||
| XM_008759324.1 | XP_008757546.1 | 7 | 1375 | |||
| XM_008759325.1 | XP_008757547.1 | 6 | 1279 | |||
| GenBank mRNA | AF375464.1 | - | 4 | 1085 | ||
| D28512.1 | - | 5 | 1187 | |||
| EU441216.1 | - | 4 | 1090 | |||

Syt3
Genomic Map