siRNA Details
Gene Symbol: St6galnac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019123.1 | NP_061996.1 | 2 | 323 | ||
GenBank mRNA | BC072501.1 | - | 2 | 160 | 7099426 | |
L29554.1 | - | 2 | 323 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: St6galnac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019123.1 | NP_061996.1 | 2 | 323 | ||
GenBank mRNA | BC072501.1 | - | 2 | 160 | 7099426 | |
L29554.1 | - | 2 | 323 |