siRNA Details
| Gene Symbol: Acp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270889.1 | NP_001257818.1 | 4 | 757 | ||
| NM_019144.2 | NP_062017.2 | 4 | 699 | |||
| XM_006242691.2 | XP_006242753.1 | 6 | 1405 | |||
| XM_006242692.2 | XP_006242754.1 | 5 | 1350 | |||
| XM_006242693.2 | XP_006242755.1 | 4 | 1216 | |||
| XM_006242694.2 | XP_006242756.1 | 4 | 653 | |||
| GenBank mRNA | BC078847.1 | - | 4 | 757 | 7110955 | |
| M76110.1 | - | 4 | 697 | |||

Acp5
Genomic Map