siRNA Details
| Gene Symbol: Strn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019148.2 | NP_062021.1 | 2 | 354 | ||
| XM_006239622.2 | XP_006239684.1 | 2 | 450 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Strn| Gene Symbol: Strn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019148.2 | NP_062021.1 | 2 | 354 | ||
| XM_006239622.2 | XP_006239684.1 | 2 | 450 | |||