siRNA Details
| Gene Symbol: Vtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019156.2 | NP_062029.2 | 2 | 233 | ||
| GenBank mRNA | AY318963.1 | - | 194 | |||
| BC105821.1 | - | 2 | 233 | 7371104 | ||
| FQ209609.1 | - | 2 | 245 | |||
| FQ209682.1 | - | 2 | 238 | |||
| FQ210518.1 | - | 2 | 238 | |||
| FQ212575.1 | - | 2 | 232 | |||
| FQ218318.1 | - | 2 | 245 | |||
| FQ219297.1 | - | 2 | 246 | |||
| U44845.1 | - | 2 | 223 | |||

Vtn
Genomic Map