siRNA Details
| Gene Symbol: Aqp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019157.2 | NP_062030.2 | 7 | 1070 | ||
| GenBank mRNA | AB000507.2 | - | 7 | 1070 | ||
| BC072695.1 | - | 7 | 1030 | 7104684 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aqp7| Gene Symbol: Aqp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019157.2 | NP_062030.2 | 7 | 1070 | ||
| GenBank mRNA | AB000507.2 | - | 7 | 1070 | ||
| BC072695.1 | - | 7 | 1030 | 7104684 | ||