siRNA Details
| Gene Symbol: Aqp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019158.2 | NP_062031.1 | 6 | 985 | ||
| XM_006230187.2 | XP_006230249.1 | 6 | 1032 | |||
| GenBank mRNA | AB005547.1 | - | 6 | 984 | ||
| AF007775.1 | - | 6 | 964 | |||
| BC081812.1 | - | 6 | 985 | 7114305 | ||

Aqp8
Genomic Map