siRNA Details
| Gene Symbol: TRAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012288.3 | NP_036420.1 | 3 | 376 | ||
| XM_011515005.2 | XP_011513307.1 | 3 | 251 | |||
| GenBank mRNA | AB621814.1 | - | 3 | 372 | ||
| AK291751.1 | - | 3 | 262 | |||
| AK311560.1 | - | 3 | 245 | |||
| BC028121.1 | - | 3 | 376 | 5169808 | ||
| D31762.1 | - | 3 | 302 | |||
| KU178488.1 | - | 3 | 227 | |||
| KU178489.1 | - | 2 | 163 | |||

TRAM2
Genomic Map