siRNA Details
| Gene Symbol: Psen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240321.2 | XP_006240383.1 | 12 | 1869 | ||
| XM_006240322.2 | XP_006240384.1 | 12 | 2122 | |||
| GenBank mRNA | BC070887.1 | - | 12 | 1795 | 7098018 | |
| FQ231303.1 | - | 12 | 1817 | |||

Psen1
Genomic Map