siRNA Details
| Gene Symbol: Chad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019164.1 | NP_062037.1 | 4 | 1387 | ||
| XM_006247186.1 | XP_006247248.1 | 3 | 3692 | |||
| GenBank mRNA | AF004953.1 | - | 4 | 1387 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chad| Gene Symbol: Chad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019164.1 | NP_062037.1 | 4 | 1387 | ||
| XM_006247186.1 | XP_006247248.1 | 3 | 3692 | |||
| GenBank mRNA | AF004953.1 | - | 4 | 1387 | ||