siRNA Details
Gene Symbol: G3BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012297.4 | NP_036429.2 | 2, 3 | 268 | ||
NM_203504.2 | NP_987100.1 | 2, 3 | 403 | |||
NM_203505.2 | NP_987101.1 | 2, 3 | 403 | |||
XM_005263382.2 | XP_005263439.1 | 2, 3 | 377 | |||
XM_005263383.3 | XP_005263440.1 | 3, 4 | 342 | |||
XM_011532441.2 | XP_011530743.1 | 2, 3 | 263 | |||
XM_017008876.1 | XP_016864365.1 | 2, 3 | 251 | |||
XM_017008877.1 | XP_016864366.1 | 2, 3 | 430 | |||
XM_017008878.1 | XP_016864367.1 | 2, 3 | 251 | |||
XM_017008879.1 | XP_016864368.1 | 2, 3 | 277 | |||
GenBank mRNA | AB014560.1 | - | 2, 3 | 223 | ||
AF051311.1 | - | 1, 2 | 103 | |||
AF053535.1 | - | 2, 3 | 298 | |||
AF145284.1 | - | 2, 3 | 298 | |||
AK291772.1 | - | 2, 3 | 333 | |||
AK291786.1 | - | 2, 3 | 238 | |||
BC011731.2 | - | 2, 3 | 321 | 4337457 | ||
BI560692.1 | - | 2, 3 | 365 | |||
CB996973.1 | - | 2, 3 | 253 | |||
CD706717.1 | - | 2, 3 | 254 | |||
DA094091.1 | - | 2, 3 | 268 | |||
DA385285.1 | - | 2, 3 | 403 | |||
DB241424.1 | - | 2, 3 | 512 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
