siRNA Details
| Gene Symbol: Gata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019185.1 | NP_062058.1 | 6 | 1833 | ||
| GenBank mRNA | L22760.1 | - | 6 | 1833 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gata6| Gene Symbol: Gata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019185.1 | NP_062058.1 | 6 | 1833 | ||
| GenBank mRNA | L22760.1 | - | 6 | 1833 | ||