siRNA Details
| Gene Symbol: Arl4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019186.1 | NP_062059.1 | 2 | 960 | ||
| XM_006240051.2 | XP_006240113.1 | 2 | 1047 | |||
| XM_006240052.2 | XP_006240114.1 | 2 | 1096 | |||
| GenBank mRNA | BC088148.1 | - | 2 | 1046 | 7373646 | |
| X77235.1 | - | 2 | 960 | |||

Arl4a
Genomic Map