siRNA Details
| Gene Symbol: Coq3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019187.1 | NP_062060.1 | 2 | 119 | ||
| GenBank mRNA | BC081811.1 | - | 284 | |||
| BC098666.1 | - | 277 | ||||
| BC126094.1 | - | 291 | 8370738 | |||
| FQ216025.1 | - | 331 | ||||
| L20427.1 | - | 2 | 119 | |||

Coq3
Genomic Map