siRNA Details
| Gene Symbol: Cd46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019190.1 | NP_062063.1 | 5 | 602 | ||
| XM_006250478.2 | XP_006250540.1 | 5 | 1585 | |||
| XM_008769849.1 | XP_008768071.1 | 5 | 1585 | |||
| XM_008769850.1 | XP_008768072.1 | 5 | 1351 | |||
| GenBank mRNA | AB010920.1 | - | 5 | 602 | ||

Cd46
Genomic Map